Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Total Annexin A2 DuoSet IC ELISA

contains the basic components required for the development of sandwich ELISAs to measure in cell lysates

$556.50 - $2280.00

Specifications

Product Type ELISA Development Kit for Intracellular Proteins
Quantity 1 Kit
Species Human
Conjugate Biotin
Assay Range 312.50 to 20000 pg/mL
View More Specs

Products 3
Catalog Number Mfr. No. Packaging Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Packaging Quantity Price Quantity & Availability  
DYC3928E
View Documents
R&D Systems™
DYC3928E
15 Plates
Each for $2,280.00
missing translation for 'QuantityLeft'
Add to Cart
 
DYC39282
View Documents
R&D Systems™
DYC39282
2 Plates
Each for $556.50
missing translation for 'QuantityLeft'
Add to Cart
 
DYC39285
View Documents
R&D Systems™
DYC39285
5 Plates
Each for $1,037.00
missing translation for 'QuantityLeft'
Add to Cart
 
Description

Description

ELISpot kits are highly sensitive, microplate-based assays for the detection of cytokine secreting cells. This kit is designed for the detection and enumeration of IFN-γ/IL-2. Complete ELISpot kits are ready-to-run and require no assay development or refinement.

The Human Total Annexin A2 DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies human Annexin A2.
Specifications

Specifications

ELISA Development Kit for Intracellular Proteins
Human
312.50 to 20000 pg/mL
ELISA
Store the unopened product at 2°C to 8°C. Do not use past expiration date.
1 Kit
Biotin
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
302
Videos
SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.