Learn More
Cole-Parmer Cole-Parmer Humidity Sponge, Regenerable; 3" X 3" bags, 40 bags/pk
Supplier: Cole-Parmer 0719313
Humidity sponges meet all drying requirements. One bag will retain 16% moisture in a container with a volume of two gallons (476 cu. in.) The sponges are non-toxic and are Class M3.5 cleanroom-compatible.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.