Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hyaluronic Acid from Bovine Vitreous Humor, MP Biomedicals™

Catalog No. MP215993310 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:

Hyaluronic Acid from Bovine Vitreous Humor, White Powder

  • It is used in study of activation of the MAPK ERK following Leishmania amazonensis, infection of Macrophages.
  • A high viscosity mucopolysaccharide composed of repeating dimeric units of glucuronic acid and N-acetyl glucosamine which forms the core of complex proteoglycan aggregates found in extracellular matrix.

Specifications

CAS

9004-61-9

Color

White

Molecular Formula

C28H44N2NaO23

MDL Number

MFCD01773053

Synonym

Poly(β-glucuronic acid-[1→3]-β-N-acetylglucosamine-[1→4]), alternating

Quantity

10mg

Solubility Information

Dissolves at 10mg/mL in analyticalgrade water togive a clear, viscous solution, after soaking overnight at +5°C then stirring at room temperature for approximately 1 hour.

InChI Key

YWIVKILSMZOHHF-QJZPQSOGSA-N

Molecular Weight (g/mol)

799.64

Physical Form

Powder

RTECSNumber

MT7250000

storageNote1

Store at +4°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.