Learn More
Thomas Scientific HyBlot ESr Autoradiography Film, HyBlot ESr High Sensitivity Film, 5 x 7"
SDP
Supplier:  Thomas Scientific 1156P38
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Extra sensitive autoradiography film, x-ray film For blotting & sequence autoradiography Use with 14C, 35S, 32P, 33P and 125I Optimized for chemiluminescence Compatible with automatic or manual processors Ultra clear, double emulsion xray film HyBlot ES® is an exceptionally clear, blue sensitive, double emulsion film that is ideal for all general purpose/autoradiography applications. It is coated on both sides to ensure maximum sensitivity and consistently produces publication quality results. Packaged 100 films per box and sealed in a light-tight envelope.
Catalog No. NC1618223