Learn More
Supply Solutions Honeywell - Hydranal Coulomat AG. Size: 1L; Grade: Reagent; CAS: 6106-24-7.; Size: 1L; Grade: Reagent; Assay: -; CAS: 6106-24-7; Hazardous: Y; Container Material: Coated Glass
Supplier:  Supply Solutions HYDRANAL 348361L
Encompass_Preferred
Coulometric Reagents!AnolytesCoulometric Reagents!Anolytes!For coulometry in cells with or without a diaphragm. Free of carbon tetrachloride and chloroform.; Size: 1L; Grade: Reagent; Assay: -; CAS: 6106-24-7; Hazardous: Y; Container Material: Coated Glass; Density: 0.930g/cm3; Compatibility reagent for coulometric KF titration (anolyte solution); Composition diethanolamine imidazole methanol; sulfur dioxide
Catalog No. NC0989800
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.