Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrobromic Acid, 47.0%, Suprapur™, MilliporeSigma™

Catalog No. p-7162451 Shop All MilliporeSigma Products
Click to view available options
Quantity:
250 mL
1 L

Chemical Identifiers

CAS 10035-10-6
Molecular Formula BrH
Molecular Weight (g/mol) 80.91
MDL Number MFCD00011323
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-N
Synonym Hydrogen bromide solution
IUPAC Name hydrogen bromide
SMILES Br

Specifications

Melting Point −11°C
Boiling Point 126°C (1013 hPa)
Density 1.49 g/cm3 (at 20°C)
Packaging Glass Bottle
Vapor Pressure 10.6 hPa (20°C)
Assay Percent Range ≥ 47%
Quantity 250 mL
Recommended Storage Store at 5 to 30°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.