Learn More
Newcomer Supply Hydrochloric Acid 5%, Aqueous, 1 Liter
Newcomer Supply Hydrochloric Acid 5%, Aqueous for Acid Cleaning Glassware is instrumental in ensuring that laboratory glassware/plasticware is acid/chemically cleaned prior to solution preparation and/or performing any silver, iron or calcium staining procedures. Shelf Life is 1 year from date of manufacture. Store at 15-30°C.