Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, 5N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog No. p-5391433 Shop All Avantor Products
Click to view available options
Quantity:
1L
4L
19L
Packaging:
Narrow Mouth Polyethylene Bottle
Poly Bottle
Poly Pail

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
SMILES [H+].[Cl-]

Specifications

IUPAC Name chlorane
Formula Weight 36.46
Grade BAKER ANALYZED™ Reagent
Packaging Narrow Mouth Polyethylene Bottle
Specific Gravity 1L = 1.08kg
Quantity 1L
Chemical Name or Material Hydrochloric Acid, 5N Volumetric Solution
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.