Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric acid solution, 0.01M, Honeywell Fluka™

Catalog No. 60046566 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:

Volumetric, 0.01 M HCl (0.01N)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

Chemical Identifiers

CAS

7647-01-0

Molecular Formula

ClH

Molecular Weight (g/mol)

36.46

MDL Number

MFCD00011324

InChI Key

VEXZGXHMUGYJMC-UHFFFAOYSA-N

Synonym

hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof

PubChem CID

313

ChEBI

CHEBI:17883

SMILES

[H+].[Cl-]

Specifications

Vapor Pressure

190 mmHg (25°C)

Quantity

6 x 1 L

UN Number

NONH for all modes of transport

IUPAC Name

hydrogen chloride

Formula Weight

36.46

Chemical Name or Material

Hydrochloric acid solution, volumetric, 0.01 M HCl (0.01N)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.