Learn More
World Precision Instrument Hydrogen Ion Exchanger
SDP
Supplier:  World Precision Instrument IE010
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Make micropipettes to record cellular concentrations
WPI’s Liquid Ion Exchangers (LIX), for use with the FD223A Electrometer, allow intracellular measurements to be made for cations (hydrogen, potassium and calcium).
Catalog No. 50-252-1175