Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hydrogen Peroxide, 30% Solution, AR, For ACS analysis, Macron Fine Chemicals™
Shop All Avantor Products
Click to view available options
Quantity:
500 mL
8 pt.
Packaging:
Plastic Bottle
Poly Container
Specifications
Specifications
Color | Colorless |
Assay Percent Range | 29.0 to 32.0 % |
Packaging | Plastic Bottle |
Quantity | 500 mL |
Formula Weight | 34.01 |
Grade | For ACS analysis |
Physical Form | Liquid |
Chemical Name or Material | Hydrogen Peroxide, 30% Solution, ACS |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction