Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More

Fisher Science Education™ Hydrogen Peroxide Solution, 3%

$8.01 - $27.15
Chemical Identifiers
CAS | 7722-84-1, 7732-18-5 |
---|---|
Molecular Formula | H2O2 |
Molecular Weight (g/mol) | 34.014 |
InChI Key | MHAJPDPJQMAIIY-UHFFFAOYSA-N |
Synonym | oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone |
PubChem CID | 784 |
ChEBI | CHEBI:16240 |
IUPAC Name | hydrogen peroxide |
SMILES | OO |
Description
This product is for educational use - no C of A is available.
Chemical Identifiers
7722-84-1, 7732-18-5 | |
34.014 | |
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
CHEBI:16240 | |
OO |
H2O2 | |
MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
784 | |
hydrogen peroxide |
Specifications
Colorless | |
H2O2 | |
1g/mL | |
MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
hydrogen peroxide | |
784 | |
34.01 | |
Hydrogen Peroxide Solution, 3% |
7722-84-1, 7732-18-5 | |
H2O2 | |
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
OO | |
34.014 | |
CHEBI:16240 | |
Liquid |
Safety and Handling

ShelfLife : 1 year
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title