Learn More
Hydrogen Potassium ATPase Beta Antibody (2G11), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 5 publications
$196.10
Specifications
Antigen | Hydrogen Potassium ATPase Beta |
---|---|
Clone | 2G11 |
Dilution | Western Blot 1:4000, Flow Cytometry 1:50, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:2000, Immunohistochemistry-Frozen 1:2000, Dot Blot 1:100 - 1:2000, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 |
Applications | Western Blot |
Classification | Monoclonal |
Description
Hydrogen Potassium ATPase Beta Monoclonal specifically detects Hydrogen Potassium ATPase Beta in Human, Mouse, Rat, Porcine, Bovine, Canine, Ferret, Rabbit samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Dot Blot.Specifications
Hydrogen Potassium ATPase Beta | |
Western Blot 1:4000, Flow Cytometry 1:50, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:2000, Immunohistochemistry-Frozen 1:2000, Dot Blot 1:100 - 1:2000, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 | |
Monoclonal | |
Ascites | |
RUO | |
ATP6B, ATPase, H+/K+ exchanging, beta polypeptide, ATPase, H+/K+ transporting, beta polypeptide, Gastric H(+)/K(+) ATPase subunit beta, gastric H+/K+ ATPase beta subunit, gastric hydrogen-potassium ATPase, beta, potassium-transporting ATPase beta chain, potassium-transporting ATPase subunit beta, Proton pump beta chain | |
ATP4B | |
IgG1 | |
Unpurified | |
Detects the beta-subunit of hydrogen/potassium ATPase. |
2G11 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
496 | |
Purified 34 kDa core peptide from deglycosylated hog gastric microsomes. | |
Primary | |
Store at -20°C. Avoid freeze/thaw cycles. |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.