Learn More
Hydrogen Potassium ATPase Beta Antibody (SR1980), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322395100UL
Description
Hydrogen Potassium ATPase Beta Monoclonal antibody specifically detects Hydrogen Potassium ATPase Beta in Human samples. It is validated for Western BlotSpecifications
Hydrogen Potassium ATPase Beta | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
SR1980 | |
Western Blot 1:500-1:2000 | |
ATP6B, ATPase, H+/K+ exchanging, beta polypeptide, ATPase, H+/K+ transporting, beta polypeptide, Gastric H(+)/K(+) ATPase subunit beta, gastric H+/K+ ATPase beta subunit, gastric hydrogen-potassium ATPase, beta, potassium-transporting ATPase beta chain, potassium-transporting ATPase subunit beta, Proton pump beta chain | |
A synthesized peptide derived from human Hydrogen Potassium ATPase Beta (Uniprot #: P51164) | |
100 μg | |
Endocrinology, Signal Transduction | |
496 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.