Learn More
Ace Glass, Inc. HYDROGENATION TRAP C
Hydrogenation gas trap with bottle and nylon bushings with o-rings is used normally between the hydrogen source and the reaction flask in Parr hydrogenation units.