Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hydroxylamine Hydrochloride, 6% w/v, Low Mercury, Reagents
Click to view available options
:
Quantity: 10 L
Specifications
Specifications
Boiling Point | 102°C |
Color | Colorless |
Physical Form | Liquid |
Quantity | 10 L |
Percent Purity | 6% |
Grade | Reagent |
Packaging | Cubitainer |
Specific Gravity | 1.0259 |
Chemical Name or Material | Hydroxylamine Hydrochloride, 6% w/v, Low Mercury |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction