Learn More
Biotium Hydroxystilbamidine
Supplier: Biotium 80014
Hydroxystilbamidine (also called Fluoro-Gold) has been used extensively as a retrograde tracer for neurons and also a histochemical stain. Fluoro-Gold can undergo retrograde axonal transport, it shows extensive filling of dendrites, and has a high resistance to fading. Also see hydroxystilbamidine 4% in H2O, which is a more convenient, ready-to-use form. Properties: Ex/Em = 361/536 nm; Yellow solid soluble in water; Store at 4°C and protect from light; C18H24N4O7S2; MW: 473; [223769-64-0].Fluoro-Gold is a trademark of Fluorochrome, LLC.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.