Learn More
Invivogen Hygromycin B Gold, Selective antibiotic for the hph gene, 31282-04-9, MFCD06795479, 5g (1x50mL)
Supplier: Invivogen ANTHG5
Molecular Formula C20 H37 N3 O13, Purity: ≥ 90% (HPLC), Molecular Weight 527.520, Hygromycin B is an aminoglycoside antibiotic produced by Streptomyces hygroscopicus. It kills bacteria, fungi and higher eukaryotic cells by inhibiting protein synthesis.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.