Learn More
AG Scientific Inc Hygromycin B in PBS, 10X2 ML
Supplier: AG Scientific Inc H1012PBS10X2ML
The concentration of Hygromycin B in PBS buffer is 50 mg/ml. Hygromycin B is an aminoglycoside antibiotic isolated from Streptomyces hygroscopicus. It is commonly used to study protein synthesis. Aside from its efficacy against bacteria, fungi, and higher eukaryotic cells, hygromycin B is also an antiviral agent inhibiting translation upon selectively entering cells made permeable due to viral infection. It works as a standard selection antibiotic in gene experiments, especially for selection of hygromycin resistance gene transformed cells. Hygromycin's longstanding application as a gene selection antibiotic, has a powerful new role in genome editing utilizing the CRISPR/CAS9 system.CAS Number: 31282-04-09Molecular Weight: 527.53Chemical Formula: C20H37N3O13Solubility: Soluble in WaterStorage Temperature: 2-8CResearch or further manufacturing use only, not for food or drug use.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.