Learn More
AdipoGen Hypericin
SDP
Supplier:  AdipoGen AGCN20449M001
Encompass_Preferred
Chemical. CAS 548-04-9. Formula C30H16O8. MW 504.5. Isolated from Hypericum perforatum. One of the principal active constituents of St. Johns Wort Hypericum perforatum. Hypericin is a photosensitive pigment/chromophore with bright red fluorescence emission lambdamax 594nm. Has a high triplet quantum yield and ability to produce considerable amounts of singlet oxygen and other ROS species when photoactivated. Antiviral, anticancer and antidepressant agent. Inactivates enveloped viruses including HIV. This photosensitizing agent has shown to induce apoptosis and necrosis in cancer cells during photodynamic therapy. Selective inhibitor of protein kinase C PKC. Has been shown to inhibit other kinases and targets such as EGFR-PTK, PI3K, CKII, MAPK and insulin receptor IR. Necrosis-avid contrast agent investigated for use in non-invasively targeting necrotic tissues, based on to be defined components in the phospholipid bilayer.
Catalog No. 50-256-2522
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.