Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ HYPERStack™ Untreated Cell Culture Vessels - New Design

Catalog No. p-7214998 Shop All Corning Products
Click to view available options
Culture Area:
18,000 cm2
6000 cm2
Product Type:
12-layer vessel
36-layer vessel
No. per Case:
2/Cs.
4/Cs.
This item is not returnable. View return policy
This item is not returnable. View return policy

More cells – provides up to 5X the growth surface area of a traditional cell culture vessel of comparable footprint.

Corning's High Yield PERformance (HYPER) Platform - Closed System for High Yield Cell Growth

The Corning HYPERStack cell culture vessel combines the best of two Corning products: the Corning CellSTACK™ and the Corning HYPERFlask™ cell culture vessels. The utilization of gas-permeable film technology provided in the spatial footprint of the CellSTACK vessel allows the HYPERStack platform to be among the most efficient, scalable cell culture vessels for adherent cell culture available today.

Features and Benefits

  • More cells – provides up to 5X the growth surface area of a traditional cell culture vessel of comparable footprint
  • Closed system – no open fluid manipulations
  • Scalable product – multiple size offerings support scale-up and scale-out
  • Ergonomic design – easier manipulation with handling equipment and accessories
  • Fixed media volume – 0.2 mL/cm2 fills vessel for less volumetric waste

Surface Treatment Guide

Corning CellBIND™ surface treatment is designed to enhance cell attachment and increase cell yields, improving attachment of loosely adherent cells and maintaining growth in reduced-serum or serum-free conditions. Alternatively, a non-treated option can support conventional cell attachment and allow for a faster and gentler harvest procedure for downstream processing and assays.

Specifications

Capacity (Metric) 1310 mL
Material Polystyrene
No. per Case 4/Cs.
No. per Pack 1 Ea.
Product Type 12-layer vessel
Shape Rectangular
Sterility Sterile
Culture Area 6000 cm2
Surface Treatment Non-treated
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.