Learn More
Revvity Health Sciences Inc HypoxiSense 680 Fluorescent Imaging Agent
Supplier: Revvity Health Sciences Inc NEV11070
Hypoxia occurs in tumors as a result of the poor ability of their disorganized vascular networks to deliver blood borne oxygen. HypoxiSense680 fluorescent imaging agent detects the tumor cell surface expression of carbonic anhydrase 9 (CA IX) protein, which increases in hypoxic regions within many tumors, especially in cervical, colorectal, non-small cell lung tumors. Pairing HypoxiSense with optical fluorescent imaging technology allows you to image and quantitate tumor sub-regions undergoing hypoxia-related changes, non-invasively and in vivo. HypoxiSense in vivo imaging reagent is ideally suited for detecting hypoxia-induced changes in CA IX expression during: Imaging in vivo mouse and rat tumors Assessing therapeutic efficacy in drug screening of tumor models Fluorescence microscopy of disease tissues For the first time, hypoxia can be measured non-invasively in living mice and rats, as well as in ex vivo tissue samples. It's the perfect solution for the in vivo hypoxia researche
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.