Learn More
ZeptoMetrix Candida albicans Z006, Titered
Shop All ZeptoMetrix Corporation Products

- Each frozen aliquot contains 1mL of pure, titered culture
- Identification of the organism was confirmed by rDNA sequencing
- Biosafety level 1 microorganism
- Should be used within the confines of a Biological Safety cabinet
IMPORTANT: Contains live material. Customer must submit an executed ZeptoMetrix MTA before vendor can ship order.
Specifications
Biosafety Level
1
Certifications/Compliance
ISO, 13485
Content And Storage
-65°C
Media Type
Vial
Product Type
Titered Live Bacteria Control
For Use With (Application)
Cross-reactivity studies, limit of detection studies, Nucleic Acid or molecular testing, other viral based assays
Quantity
1 mL
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.