Learn More
Invitrogen™ Ambion™ mirVana™ miRNA Inhibitor, let-7c positive control

miRNA inhibitors are small, chemically modified single-stranded RNA molecules designed to specifically bind to and inhibit endogenous miRNA molecules and enable miRNA functional analysis by down-regulation of miRNA activity.
$365.00 - $1058.00
Specifications
Content And Storage | Store at or below -20°C. Do not store in a frost-free freezer. |
---|---|
Control Type | Positive Control |
RNAi Type | miRNA Inhibitor |
For Use With (Application) | miRNA & Non-Coding RNA Analysis |
Label or Dye | Unconjugated |
Description
miRNA inhibitors are small, chemically modified single-stranded RNA molecules designed to specifically bind to and inhibit endogenous miRNA molecules and enable miRNA functional analysis by down-regulation of miRNA activity.
We have now released the second generation of miRNA inhibitors, mirVana™ miRNA Inhibitors, which:
- Display better potency than lead competitor's product
- Offer maximum and consistent effect in vitro at low miRNA inhibitor concentration
- Target the most updated Sanger content
- Can be used in vitro and in vivo, offering consistency throughout your entire research project
- Are non-toxic
Positive and negative controls are also available for use along with mirVana™ miRNA Inhibitors in miRNA down-regulation experiments.
mirVana™ miRNA Inhibitor let-7c Positive Control
The mirVana™ miRNA Inhibitor let-7c Positive Control is intended for use as a positive control for mammalian cell culture experiments using mirVana™ miRNA Inhibitors. Endogenous let-7c miRNA has been shown to negatively regulate HMGA2 mRNA in cultured cells. HMGA2 is a ubiquitously expressed, nonhistone, chromatin protein that can modulate gene expression through changes in chromatin architecture. let-7c miRNA down-regulates levels of HMGA2 mRNA. When transfected into human and mouse cell lines, let-7c miRNA inhibitor blocks endogenous let-7c miRNA, resulting in increased levels of HMGA2 mRNA. Thus, let-7c miRNA inhibitor activity can be monitored in human or mouse cells using real-time reverse transcription PCR (RT-PCR) to detect HMGA2 mRNA.
First Generation Inhibitor Controls
Our first-generation miRNA inhibitors (and controls), Ambion™ Anti-miR™ miRNA Inhibitors, have lower specificity and potency, plus a lower strand specificity compared to the new mirVana™ miRNA Inhibitors. These first-generation inhibitors are recommended for completing ongoing in vitro miRNA studies. For new studies or in vivo studies, we highly recommend our new mirVana™ miRNA Inhibitors and their corresponding controls.
Specifications
Store at or below -20°C. Do not store in a frost-free freezer. | |
miRNA Inhibitor | |
Unconjugated | |
Purified |
Positive Control | |
miRNA & Non-Coding RNA Analysis | |
miRNA Inhibitor | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.