Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Phosphoric Acid, 85.0% W/W, MilliporeSigma™

Catalog No. p-7162442 Shop All MilliporeSigma Products
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Glass Bottle, Pre-Packed
Plastic Bottle

Chemical Identifiers

CAS 7664-38-2
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.99
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
IUPAC Name phosphoric acid
SMILES OP(O)(O)=O

Specifications

pH < 0.5 (100 g/L, H2O, 20°C)
Melting Point 21°C
Boiling Point 158°C (1013 mbar)
Packaging Glass Bottle, Pre-Packed
Vapor Pressure 2 hPa (20°C)
Assay Percent Range 85.0% min
Quantity 500 mL
Recommended Storage Store at room temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.