Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Pseudomonas putida ATCC™ 31483 ™†
SDPSCI_ED

For culture purposes and QC applications.

Supplier:  Microbiologics™ 0702P/EMD

Catalog No. S3-591-3

This item is not returnable. View return policy

Description

Description

KWIK-STIK™
  • Clever design and accurate, repeatable results
  • Each device features a single Pseudomonas putida strain in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
  • For added convenience, a peel-off identification label is included for easy documentation
  • Available in pack of 2
  • ATCC™ 31483 ™†
Specifications

Specifications

31483
CE, online certificate of analysis
Lyophilized Pellet
Pseudomonas putida QC Microorganism
Culture purpose and QC application
1
2°C to 8°C
Kwik-stik™
Pseudomonas putida QC microorganism
2 U
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 6 to 24 Months

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.