Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl Alcohol, ACS, 99.0%, MilliporeSigma™

Catalog No. p-7162443 Shop All MilliporeSigma Products
Click to view available options
Packaging:
Plastic Bottle
Tinplate Metal Can

Chemical Identifiers

CAS 75-65-0
Molecular Weight (g/mol) 74.12

Specifications

Melting Point 24 to 25°C
Boiling Point 81°C to 83°C (1013 hPa)
Quantity 1 L
Flash Point 14°C
Packaging Plastic Bottle
Residue after Evaporation 0.003% max
Assay Percent Range 99.0% min.
Vapor Pressure 40.7 hPa (20°C)
Recommended Storage Store at room temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.