Learn More
Inorganic Ventures 1000ug mL CALCIUM 125mL (Inorganic Ventures)
SDP
Supplier:  Inorganic Ventures ICCA1125ML
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Inorganic Ventures' 1000 ppm (g/mL) Calcium for IC is a 125mL certified reference material set in a nitric acid matrix for stability. This calcium standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. One year Expiration Date.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-555