Learn More
Medchemexpress LLC HY-101396 5mg Medchemexpress, ICA-069673 CAS:582323-16-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1013965MG
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Medchemexpress, HY-101396 5mg ICA-069673 CAS:582323-16-8 ICA-069673 is a KCNQ2/Q3 potassium channel activator with an IC 50 of 0.69 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2107
missing translation for 'QuantityLeft'