Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ICI 118,551 hydrochloride, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
ICI 118,551 hydrochloride is a very selective β2 adrenergic inverse agonist (Ki values are 1.2, 120 and 257 nM for β2, β1 and β3 receptors respectively). Active in vivo.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 313.87 |
Percent Purity | >98% |
Physical Form | Solid |
Chemical Name or Material | ICI 118,551 hydrochloride |
Safety and Handling
Recommended Storage | Store at room temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction