Learn More
Inorganic Ventures 1000ug mL CALCIUM 125mL (Inorganic Ventures)
SDP
Supplier:  Inorganic Ventures CGCA1125ML
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Matrix: Nitric Acid (HNO3), Starting Material: CaO, Analyte: Ca 1000 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-228
missing translation for 'QuantityLeft'