Learn More
Inorganic Ventures 1000ug mL CARBON 125mL (Inorganic Ventures)
Supplier: Inorganic Ventures CGC1125ML
Inorganic Ventures' 1000 ppm (g/mL) Carbon for ICP is a 125mL certified reference material set in a nitric acid matrix for stability. This carbon standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. One year Expiration Date.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.