Learn More
Inorganic Ventures 1000ug mL CARBON 500mL (Inorganic Ventures)
SDP
Supplier:  Inorganic Ventures CGC1500ML
Encompass_Preferred
Inorganic Ventures' 1000 ppm (g/mL) Carbon for ICP is a 500mL certified reference material set in a nitric acid matrix for stability. This carbon standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. One year Expiration Date.
Catalog No. 50-723-225
missing translation for 'QuantityLeft'
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.