Learn More
Inorganic Ventures 100ug mL COPPER 125mL (Inorganic Ventures)
SDP
Supplier:  Inorganic Ventures MSCU100PPM125ML
Encompass_Preferred
Matrix: Nitric Acid (HNO3), Starting Material: Cu metal, Analyte: Cu 100 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-725
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.