Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

iCRT 14, Tocris Bioscience™

Catalog No. p-4817051 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Inhibits β-catenin-responsive transcription (CRT)

Potent inhibitor of β-catenin-responsive transcription (CRT) (IC50 = 40.3 nM in assays of Wnt pathway activity). Thought to directly influence the interaction between β-catenin and TCF4. Induces marked G0/G1 cell cycle arrest in HCT-116 and HT29 cell lines.

Chemical Identifiers

CAS 677331-12-3
Molecular Formula C21H17N3O2S
Molecular Weight (g/mol) 375.446
InChI Key NCSHZXNGQYSKLR-UNOMPAQXSA-N
Synonym icrt 14, icrt-14, icrt14, 5-2,5-dimethyl-1-3-pyridinyl-1h-pyrrol-3-yl methylene-3-phenyl-2,4-thiazolidinedione, 5z-5-2,5-dimethyl-1-pyridin-3-ylpyrrol-3-yl methylidene-3-phenyl-1,3-thiazolidine-2,4-dione, icrt hplc, 5-2,5-dimethyl-1-3-pyridinyl-1h-pyrrol-3-yl methylene-3-phenyl-2,4-thiazolidinedion, 5z-5-2,5-dimethyl-1-pyridin-3-yl-1h-pyrrol-3-yl methylidene-3-phenyl-1,3-thiazolidine-2,4-dione, 5z-5-2,5-dimethyl-1-pyridin-3-yl pyrrol-3-yl methylidene-3-phenyl-1,3-thiazolidine-2,4-dione
PubChem CID 5967294
IUPAC Name (5Z)-5-[(2,5-dimethyl-1-pyridin-3-ylpyrrol-3-yl)methylidene]-3-phenyl-1,3-thiazolidine-2,4-dione
SMILES CC1=CC(=C(N1C2=CN=CC=C2)C)C=C3C(=O)N(C(=O)S3)C4=CC=CC=C4

Specifications

Quantity 10 mg
Formula Weight 375.44
Percent Purity >99%
Chemical Name or Material iCRT 14
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.