Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IDE 1, Tocris Bioscience™

Catalog No. p-4816829 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Induces definitive endoderm formation in mouse and human ESCs

Induces definitive endoderm formation in mouse and human embryonic stem cells (ESCs) (EC50 = 125 nM). Thought to activate the TGF-β signaling pathway; induces Smad2 phosphorylation and increases levels of Nodal expression.

This compound is mixture of E/Z isomers.

Chemical Identifiers

CAS 1160927-48-9
Molecular Formula C15H18N2O5
Molecular Weight (g/mol) 306.318
InChI Key ABKJCDILEUEJSH-UHFFFAOYSA-N
Synonym ide 1
PubChem CID 66974738
IUPAC Name 2-[(6-carboxyhexanoylhydrazinylidene)methyl]benzoic acid
SMILES C1=CC=C(C(=C1)C=NNC(=O)CCCCCC(=O)O)C(=O)O

Specifications

Quantity 10 mg
Formula Weight 306.31
Percent Purity >98%
Chemical Name or Material IDE 1
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.