Learn More
Revvity Health Sciences Inc IgM (bovine) AlphaLISA Detection Kit, 5,000 Assay Points
Supplier: Revvity Health Sciences Inc AL533F
Formats: Our HV (100 assay point) kits allow you to run 100 wells in 96-well format, using a 100 ¿L reaction volume (10 ¿L of sample). Our 500 assay point kit allows you to run 500 wells in 96-well or 384-well format, using a 50 ¿L reaction volume (5 ¿L of sample). Our 5,000 assay point kit allows you to run 5,000 wells in 96-well or 384-well format, using a 50 ¿L reaction volume (5 ¿L of sample). Features: No-wash steps, no separation steps ELISA alternative technology Sensitive detection Broad sample compatibility Small sample volume Results in less than 3 hours Half the time of an ELISA assay There are five classes of mammalian immunoglobulins: IgA, IgD, IgE, IgM, and IgG. IgG is the most abundant immunoglobulin and is equally distributed in blood and tissue. The general immunoglobulin structure is composed of four polypeptide chains, two heavy and two light chains linked together and to each other by disulfide bonds, creating a tetrameric quaternary structure. Bovine immunoglobul
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.