Learn More
IgM Fc fragment Porcine anti-Human, Polyclonal, Secondary Antibody, Novus Biologicals™

Pig Polyclonal Antibody
Supplier: Novus Biologicals NBP269715
Description
IgM Fc Polyclonal specifically detects IgM Fc in Human samples. It is validated for Immunoprecipitation.Specifications
IgM Fc fragment | |
LYOPH | |
Immunoprecipitation 1:10 - 1:500 | |
Antiserum, Lyophilized | |
Highly purified normal IgM isolated from pooled human serum. Freund's complete adjuvant is used in the first step of the immunization procedure. | |
1 mL | |
Secondary | |
Reconstitute the lyophilized antiserum by adding 1 ml sterile distilled water. Dilutions may be prepared by adding phosphate buffered saline (PBS, pH 7.2). |
Immunoprecipitation | |
Unconjugated | |
Whole antiserum with No Preservative | |
Porcine | |
Whole Antiserum | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
Polyclonal | |
Human |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.