Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IKK 16, Tocris Bioscience™

Catalog No. p-4784032 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective inhibitor of IKK

Selective inhibitor of IκB kinase (IKK) (IC50 values are 40, 70 and 200 nM for IKKβ, IKK complex and IKKα respectively). Inhibits TNF-α-stimulated IκB degradation and expression of adhesion molecules E-selectin, ICAM and VCAM (IC50 values are 1.0, 0.5, 0.3 and 0.3 μM respectively). Inhibits LPS-induced TNF-α release in vivo and neutrophil extravasion in thioglycollate-induced peritonitis.

Chemical Identifiers

CAS 1186195-62-9
Molecular Formula C28H30ClN5OS
Molecular Weight (g/mol) 520.092
InChI Key XFVDIVQCZWDBCW-UHFFFAOYSA-N
Synonym ikk 16, ikk-16, ikk-16 hcl, 4-1-benzothiophen-2-yl-n-4-4-pyrrolidin-1-yl piperidine-1-carbonyl phenyl pyrimidin-2-amine hydrochloride
PubChem CID 56972179
IUPAC Name [4-[[4-(1-benzothiophen-2-yl)pyrimidin-2-yl]amino]phenyl]-(4-pyrrolidin-1-ylpiperidin-1-yl)methanone;hydrochloride
SMILES C1CCN(C1)C2CCN(CC2)C(=O)C3=CC=C(C=C3)NC4=NC=CC(=N4)C5=CC6=CC=CC=C6S5.Cl

Specifications

Quantity 10 mg
Formula Weight 520.09
Percent Purity >98%
Chemical Name or Material IKK 16
Recommended Storage Desiccate at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.