Learn More
Selleck Chemical LLC Iloperidone 10mg 133454-47-4 HP873
Supplier:  Selleck Chemical LLC S148310MG
Encompass_Preferred
Iloperidone (HP873) is a dopamine (D2)/serotonin (5-HT2) receptor antagonist, used for the treatment of schizophrenia. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-733-5
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.