Learn More
Invitrogen™ Image-iT™ Plant Tissue Hard-Set Mountant

Image-iT Plant Tissue Hard-Set Mountant is used for imaging, storage, and archiving of plant tissues optically cleared with Image-iT Plant Tissue Clearing Reagent.
$212.00
Specifications
Content And Storage | Store at 15°C to 25°C. Do not freeze. When stored as directed, product is stable for (at least 6 months after receipt. ) |
---|---|
Product Line | Image-iT™ |
Shipping Condition | Room Temperature |
Description
These reagents are compatible with both fluorescence-based and colorimetric-based applications. Image-iT Plant Tissue mountant is based on Visikol™ Mount, which has a history of use in fluorescent and colorimetric studies with plants.
• Clears and mounts plant tissue for whole specimen imaging
• Hard-set mountant with refractive index of 1.415
Specifications
Store at 15°C to 25°C. Do not freeze. When stored as directed, product is stable for (at least 6 months after receipt. ) |
|
Image-iT™ | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.