Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Three Quarter Finger Anti-Impact Nylon Lycra™ Gloves with VEP Padding

Catalog No. 19039282 Shop All Impacto Protective Products Products
Click to view available options
:
This item is not returnable. View return policy
This item is not returnable. View return policy

VEP padding with a grain leather cover on the palm provides protection from impact and abrasion. Nylon fabric on the back ensures breathability and comfort.

  • Nylon fabric ensures breathability and comfort
  • Contoured VEP padding in the palm provides protection from impact and shock
  • 3/4 finger design leaves finger tips open to for tactile feedback and mobility
  • Grain leather extends glove's useful life

Specifications

Back Material

Nylon Lycra™

Color

Blue

Hand

Specific

Material

Grain Leather

Size

Large

For Use With (Application)

Construction, Assembly, Manufacturing, Maintenance, and Warehousing

Glove Cuff Style

Open

Series

505-20

Length (English)

9 in.

Length (Metric)

23 cm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.