Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ 409-30 Half-Finger Trigger Gloves without Wrist Support

Catalog No. 189994596D Shop All Impacto Protective Products Products
Click to view available options
Size:
Hand:

Gloves are constructed of soft pearl leather on the side of the palm, with a stretchy Nylon Lycra™ back. VEP padding in the palm, trigger finger, and thumb web area provides additional impact protection.

  • Soft pearl leather palm offers dexterity and abrasion resistance
  • Nylon Lycra™ back ensures breathability and comfort
  • VEP 1/8 in. padding in the palm, thumb, and index finger to protect from impact and shock
  • Half finger design with extended index finger ensures tactile feedback and mobility
  • Elastic wrist with an adjustable hook and loop closure
  • Pull tabs at the fingertips for easy removal
  • Available with leather wrist support as 479-31

Specifications

Back Material

Nylon Lycra™

Certifications/Compliance (Europe)

EN 420: 2003 + A1:2009

Cuff Material

Elastic

Color

Black, White

Hand

Left

Size

X-Large

EN388 Standard

EN388:2222

Finger Style

Half

For Use With (Application)

Construction, Manufacturing, and Maintenance

Glove Cuff Style

Hook and Loop

Palm Type

Pearl Leather

Series

409-30

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.