Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Fingerless Anti-Impact Nylon Gloves with Detachable Wrist Support

Catalog No. 19031840 Shop All Impacto Protective Products Products
Click to view available options
:
This item is not returnable. View return policy
This item is not returnable. View return policy

A grain leather palm offers durability and abrasion resistance, while detachable wrist support protects from impact and repetitive strain injuries.

  • Stretch Nylon Lycra™ material offers breathability and comfort
  • Contoured VEP 1/8 in. padding in the palm protects from impact
  • Fingerless style for tactile feedback and mobility
  • Grain leather offers abrasion protection and durability
  • Detachable leather wrist support prevents over-flexion and over extension to prevent repetitive strain injuries

Specifications

Cuff Material

Grain Leather

Color

Blue, Yellow

Hand

Specific

Material

Nylon Lycra

Product Type

Anti-impact Gloves

Size

Medium

Finger Style

Fingerless

For Use With (Application)

Construction, Assembly, Manufacturing, and Maintenance

Glove Cuff Color

Yellow

Series

703-20

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.