Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Half-Finger Anti-Impact Gloves with Pearl Leather Wrist Support

Catalog No. p-3668098 Shop All Impacto Protective Products Products
Click to view available options
Size:
Small
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Nylon gloves with soft pearl leather provide comfort, dexterity, and abrasion protection. The attached wrist support prevents over-extension and over-flexion, a leading cause of repetitive strain injuries.

  • Nylon fabric back ensures optimum breathability and comfort
  • Palm made of soft pearl leather for dexterity and abrasion protection
  • 1/8 in. VEP padding in the palm protects from impact and shock
  • Half finger design leaves fingers open for tactile feedback and mobility
  • Attached leather wrist support prevents over extension and over flexion to protect from RSI
  • Protects from electric, manual, and power tools
  • Use in applications such as construction, manufacturing, and automotive

Specifications

Back Material Nylon
Color Black, White
Hand Specific
Material Pearl Leather
Product Type Anti-impact Gloves
Size Small
For Use With (Application) Construction, Manufacturing, and Automotive Assembly
Glove Cuff Color White
Series 471-30
Length (English) 7 in.
Length (Metric) 18 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.