Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Three-Quarter Nylon Glove Liners with VEP Padding

Catalog No. p-3667758 Shop All Impacto Protective Products Products
Click to view available options
Size:
Small
Large
Hand:
Left
Right
This item is not returnable. View return policy
This item is not returnable. View return policy

Nylon Lycra™ glove liners feature VEP padding in the palm and fingers, with an open-finger design to ensure tactile feedback. Wear under work gloves for additional shock and impact protection.

  • Three-quarter finger design ensures tactile feedback and dexterity
  • VEP padding in the palm and fingers
  • Wear under work gloves for additional protection from shock and impact

Specifications

Color Blue
Hand Left
Material Nylon Lycra
Product Type Anti-Impact Glove Liners
Size Small
Finger Style Three Quarter
For Use With (Application) Impact and Shock Protection
Glove Cuff Style Open
Palm Type Padded
Series 505-00
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.