Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Fingerless Polycotton Anti-Impact Glove Liners with Thumb Web Padding

Catalog No. p-3667755 Shop All Impacto Protective Products Products
Click to view available options
Size:
Large
X-Large
Hand:
Left
Right
Specific
This item is not returnable. View return policy
This item is not returnable. View return policy

Designed to be worn under work gloves, anti-impact glove liners feature VEP padding in the palm and thumb web area for protection from electric, manual, and power tools.

  • 4-way stretch polycotton fabric ensures breathability and comfort
  • Contoured VEP padding in the palm and thumb web area
  • Fingerless style enhances tactile feedback and mobility
  • Designed to be worn under work gloves for additional protection

Specifications

Color Blue
Hand Left
Material Polycotton
Product Type Anti-Impact Glove Liners
Size Large
Finger Style Fingerless
For Use With (Application) Impact and Shock Protection
For Use With (Equipment) Electric, Manual, and Power Tools
Glove Cuff Style Open
Palm Type Padded
Series 502-00
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.