Learn More
MilliporeSigma™ Supelco™ In-Line Pressure Regulator, Economy Model
An inexpensive version for general lab applications
Supplier:  MilliporeSigma™ Supelco™ 23833U
 View more versions of this product
Catalog No. 11-101-1407
Add to Cart
Description
Description
Specifications
Specifications
Pneumatic Control, and Other Non-Chromatographic Purposes
1/8 in. Brass Swagelok Nuts and Ferrules
0 to 300 psi (Inlet)
Gauge Only
Gas Streams
Economy
1 Ea.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.