Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Indicator Solution, Universal
SCI_ED

Catalog No. p-4416718 Shop All Fisherbrand Products
Click to view available options
Quantity:
1 L
100 mL
500 mL
Ea.
Packaging:
Color Chart
Solution
Solution with Color Chart
This item is not returnable. View return policy
This item is not returnable. View return policy
pH indicator; for pH determinations from pH 4.0 to 10.0

This product is for educational use - no C of A is available.

Specifications

Chemical Name or Material Universal Indicator Solution
CAS 34722-90-2, 64-17-5, 547-58-0, 845-10-3, 518-51-4
Assay Percent Range <0.2% w/v
Synonym Universal pH Indicator
Packaging Solution
Density 0.93
Color Green
Quantity 100 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.