Learn More
Medchemexpress LLC HY-13650 5mg Medchemexpress, Indisulam CAS:165668-41-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY136505MG
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Medchemexpress, HY-13650 5mg Indisulam CAS:165668-41-7 Indisulam (E 7070) is a carbonic anhydrase inhibitor with anticancer activity. Indisulam (E 7070) is a sulfonamide agent that targets the G1 phase of the cell cycle. Indisulam (E 7070) causes a blockade in the G1/S transition through inhibition of the activation of both CDK2 and cyclin E . Indisulam (E 7070) targets splicing by inducing RBM39 degradation via recruitment to DCAF15 [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2674
missing translation for 'QuantityLeft'